(S)-Methyl-2-amino-6-(((benzyloxy)carbonyl)amino)hexanoate hydrochloride 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
H-Lys(Z)-OMe.HCl, H-Lys(Z)-OMe·HCl
A555611-100G
A555611-10G
A555611-1G
A555611-5G
A555611-25G
77005-116EA
283.27
USD
77005-116
77005-118
77005-120
77005-122
77572-758
(S)-Methyl-2-amino-6-(((benzyloxy)carbonyl)amino)hexanoate hydrochloride 98%
(S)-Methyl-2-amino-6-(((benzyloxy)carbonyl)amino)hexanoate hydrochloride
Precaución: for Research Use Only
Formula:
C₁₅H₂₃ClN₂O₄ MW: 330.809 g/mol |
MDL Number:
MFCD00034846 CAS Number: 27894-50-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...