(R)-N-Boc-2-aminocyclohexanone 97%, Ambeed.Inc

Supplier: Ambeed
Warning

A555445-1G A555445-250MG
76991-714EA 227.04 USD
76991-714 77684-399
(R)-N-Boc-2-aminocyclohexanone 97%, Ambeed.Inc
(R)-N-Boc-2-aminocyclohexanone

Precaución: For research use only.

Formula: C₁₁H₁₉NO₃
MW: 213.27 g/mol
MDL Number: MFCD09838824
CAS Number: 149524-64-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR