[1,3-Bis(diphenylphosphino)propane]palladium(II) dichloride 98%

Supplier: Ambeed
Warning

Synonyms: Dichloro[1,3-bis(diphenylphosphino)propane]palladium(II)

A555253-1G A555253-25G A555253-5G A555253-250MG
77034-872EA 42 USD
77034-872 77669-460 77669-462 77872-931
[1,3-Bis(diphenylphosphino)propane]palladium(II) dichloride 98%
[1,3-Bis(diphenylphosphino)propane]palladium(II) dichloride

Precaución: for Research Use Only

Formula: C₂₇H₂₆Cl₂P₂Pd
MW: 589.77 g/mol
MDL Number: MFCD03844773
CAS Number: 59831-02-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR