(S)-3-(4-Hydroxyphenyl)hex-4-ynoic acid 95%

Supplier: Ambeed
Warning

A554808-100MG A554808-250MG
76991-710EA 50.93 USD
76991-710 76991-712
(S)-3-(4-Hydroxyphenyl)hex-4-ynoic acid 95%
(S)-3-(4-Hydroxyphenyl)hex-4-ynoic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₂O₃
MW: 204.22 g/mol
MDL Number: MFCD23106483
CAS Number: 865233-35-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR