PF-562271 besylate 98%

Supplier: Ambeed
Warning

Synonyms: N-Methyl-N-(3-(((2-((2-oxoindolin-5-yl)amino)-5-(trifluoromethyl)pyrimidin-4-yl)amino)methyl)pyridin-2-yl)methanesulphonamide benzenesulphonate, N-[3-[[[2-[(2,3-dihydro-2-oxo-1H-indol-5-yl)amino]-5-(trifluoromethyl)-4-pyrimidinyl]amino]methyl]-2-pyridinyl]-N-methyl-methanesulphonamide, monobenzenesulphonate

A554072-10MG A554072-1MG A554072-2MG A554072-50MG A554072-5MG A554072-100MG
77017-696EA 53 USD
77017-696 77017-698 77017-700 77017-702 77017-704 77893-127
PF-562271 besylate 98%
PF-562271 besylate

Precaución: for Research Use Only

Formula: C₂₁H₂₀F₃N₇O₃S.CH₆O₃S
MW: 665.66 g/mol
MDL Number: MFCD14105612
CAS Number: 939791-38-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR