1-Boc-4-(2-formylphenyl)piperazine 95%

Supplier: Ambeed
Warning

Synonyms: 1-Boc-4-(2-Formylphenyl)piperazine

A553853-10G A553853-1G A553853-250MG A553853-25G A553853-5G
77034-858EA 168.69 USD
77034-858 77034-860 77034-862 77034-864 77034-866
1-Boc-4-(2-formylphenyl)piperazine 95%
1-Boc-4-(2-formylphenyl)piperazine

Precaución: for Research Use Only

Formula: C₁₆H₂₂N₂O₃
MW: 290.36 g/mol
MDL Number: MFCD05864664
CAS Number: 174855-57-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR