1,10-Phenanthroline-5-carbaldehyde 95%

Supplier: Ambeed
Warning

A552778-100MG A552778-1G A552778-250MG
77034-852EA 34 USD
77034-852 77034-854 77034-856
1,10-Phenanthroline-5-carbaldehyde 95%
1,10-Phenanthroline-5-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₃H₈N₂O
MDL Number: MFCD20528237
CAS Number: 91804-75-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR