2-Hydroxy-2-methylbutyric acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-Hydroxy-2-methylbutanoic acid

A552647-100G A552647-25G A552647-5G
76991-700EA 218.55 USD
76991-700 76991-702 76991-704
2-Hydroxy-2-methylbutyric acid 98%
2-Hydroxy-2-methylbutyric acid

Precaución: for Research Use Only

Formula: C₅H₁₀O₃
MW: 118.13 g/mol
MDL Number: MFCD00004476
CAS Number: 3739-30-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR