4-Chloro-2-fluorobenzylamine hydrochloride 98%

Supplier: Ambeed
Warning

A552584-1G A552584-250MG
77005-094EA 9 USD
77005-094 77005-096
4-Chloro-2-fluorobenzylamine hydrochloride 98%
4-Chloro-2-fluorobenzylamine hydrochloride

Precaución: for Research Use Only

Formula: C₇H₇ClFN·HCl
MDL Number: MFCD00143286
CAS Number: 202982-63-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR