(2-Amino-3-nitrophenyl)methanol 95%

Supplier: Ambeed
Danger

A552516-1G A552516-250MG
76991-696EA 58 USD
76991-696 76991-698
(2-Amino-3-nitrophenyl)methanol 95%
(2-Amino-3-nitrophenyl)methanol

Precaución: for Research Use Only

Formula: C₇H₈N₂O₃
MDL Number: MFCD09880172
CAS Number: 139743-08-1
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR