Spiro[cyclohexane-1,3'-indolin]-2'-one 95%

Supplier: Ambeed
Warning

A552445-100MG A552445-1G A552445-250MG
77017-674EA 51 USD
77017-674 77017-676 77017-678
Spiro[cyclohexane-1,3'-indolin]-2'-one 95%
Spiro[cyclohexane-1,3'-indolin]-2'-one

Precaución: for Research Use Only

Formula: C₁₃H₁₅NO
MDL Number: MFCD11935809
CAS Number: 4933-14-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR