(1-Formylcyclopropyl)carbamic acid-tert-butyl ester 98%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 1-formylcyclopropylcarbamate, 1(tert-Butoxycarbonylamino)cyclopropanecarboxaldehyde

A552266-250MG A552266-5G A552266-1G
77034-840EA 10 USD
77034-840 77034-842 77872-927
(1-Formylcyclopropyl)carbamic acid-tert-butyl ester 98%
(1-Formylcyclopropyl)carbamic acid-tert-butyl ester

Precaución: for Research Use Only

Formula: C₉H₁₅NO₃
MDL Number: MFCD10697899
CAS Number: 107259-06-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR