5-[4-(Trifluoromethyl)phenyl]-1H-tetrazole 98%

Supplier: Ambeed
Danger

A552154-250MG A552154-1G A552154-5G
77017-672EA 20 USD
77017-672 77893-124 77893-125
5-[4-(Trifluoromethyl)phenyl]-1H-tetrazole 98%
5-[4-(Trifluoromethyl)phenyl]-1H-tetrazole

Precaución: for Research Use Only

Formula: C₈H₅F₃N₄
MDL Number: MFCD06248886
CAS Number: 2251-79-8
UN: 1325
ADR: 4.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR