2-Methyl-3-tetrahydrothiophenone 97%

Supplier: Ambeed
Warning

Synonyms: Dihydro-2-methyl-3(2H)-thiophenone, 2-Methyl-3-oxotetrahydrothiophene

A552092-1G A552092-5G
77005-090EA 7.43 USD
77005-090 77005-092
2-Methyl-3-tetrahydrothiophenone 97%
2-Methyl-3-tetrahydrothiophenone

Precaución: for Research Use Only

Formula: C₅H₈OS
MW: 116.18 g/mol
MDL Number: MFCD00078280
CAS Number: 13679-85-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR