1-(2-Tetrahydropyranyl)-1H-imidazole-5-boronic acid pinacol ester 95%

Supplier: Ambeed
Warning

Synonyms: 1-(Tetrahydro-2H-pyran-2-yl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-imidazole

A550853-100MG A550853-1G A550853-250MG A550853-5G
77034-832EA 25 USD
77034-832 77034-834 77034-836 77034-838
1-(2-Tetrahydropyranyl)-1H-imidazole-5-boronic acid pinacol ester 95%
1-(2-Tetrahydropyranyl)-1H-imidazole-5-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₄H₂₃BN₂O₃
MDL Number: MFCD11100960
CAS Number: 1029684-37-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR