(1S)-2,2'-Dibromo-1,1'-binaphthalene ≥98%, ee 99%

Supplier: Ambeed
Warning

A550649-100MG A550649-250MG A550649-50MG
77005-086EA 161.26 USD
77005-086 77602-326 77673-466
(1S)-2,2'-Dibromo-1,1'-binaphthalene ≥98%, ee 99%
(1S)-2,2'-Dibromo-1,1'-binaphthalene

Precaución: for Research Use Only

Formula: C₂₀H₁₂Br₂
MW: 409.93 g/mol
CAS Number: 150024-49-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR