5-Chloroquinazoline 95%

Supplier: Ambeed
Warning

A550493-1G A550493-250MG
76991-692EA 84.88 USD
76991-692 76991-694
5-Chloroquinazoline 95%
5-Chloroquinazoline

Precaución: for Research Use Only

Formula: C₈H₅ClN₂
MW: 164.59 g/mol
MDL Number: MFCD18448930
CAS Number: 7556-90-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR