1-Bromo-2-fluoro-4-nitro-5-(trifluoromethyl)benzene 97+%

Supplier: Ambeed
Warning

A550215-10G A550215-1G A550215-250MG A550215-5G
76991-684EA 161.26 USD
76991-684 76991-686 76991-688 76991-690
1-Bromo-2-fluoro-4-nitro-5-(trifluoromethyl)benzene 97+%
1-Bromo-2-fluoro-4-nitro-5-(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₇H₂BrF₄NO₂
MW: 287.994 g/mol
MDL Number: MFCD12032178
CAS Number: 1121586-27-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR