Pyritinol 98%

Supplier: Ambeed
Warning

Synonyms: 5,5'-(Disulphanediylbis(methylene))bis(4-(hydroxymethyl)-2-methylpyridin-3-ol), Pyrithioxin, Pyridoxine Disulphide

A549996-100MG A549996-1MG A549996-25MG A549996-1G A549996-250MG
77005-074EA 58 USD
77005-074 77005-076 77005-078 77882-328 77882-329
Pyritinol 98%
Pyritinol

Precaución: for Research Use Only

Formula: C₁₆H₂₀N₂O₄S₂
MW: 368.47 g/mol
MDL Number: MFCD00151477
CAS Number: 1098-97-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR