Methyl 3-(2-cyanopropan-2-yl)benzoate 95%

Supplier: Ambeed
Warning

A549593-100MG A549593-1G A549593-250MG
77017-654EA 30.77 USD
77017-654 77017-656 77017-658
Methyl 3-(2-cyanopropan-2-yl)benzoate 95%
Methyl 3-(2-cyanopropan-2-yl)benzoate

Precaución: for Research Use Only

Formula: C₁₂H₁₃NO₂
MW: 203.23 g/mol
MDL Number: MFCD19694850
CAS Number: 872091-85-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR