2,5-Bis(2-ethylhexyl)-3,6-di(2-thienyl)-2,5-dihydropyrrolo[3,4-c]pyrrole-1,4-dione 95%

Supplier: Ambeed
Warning

A549490-100MG A549490-250MG
76991-674EA 31 USD
76991-674 76991-676
2,5-Bis(2-ethylhexyl)-3,6-di(2-thienyl)-2,5-dihydropyrrolo[3,4-c]pyrrole-1,4-dione 95%
2,5-Bis(2-ethylhexyl)-3,6-di(2-thienyl)-2,5-dihydropyrrolo[3,4-c]pyrrole-1,4-dione

Precaución: for Research Use Only

Formula: C₃₀H₄₀N₂O₂S₂
MDL Number: MFCD22200076
CAS Number: 1185885-86-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR