N-(6-(3-(tert-Butyl)-5-(2,4-dioxo-3,4-dihydropyrimidin-1(2H)-yl)-2-methoxyphenyl)naphthalen-2-yl)methanesulfonamide 98%, Ambeed.Inc

Supplier: Ambeed
Warning

Synonyms: Dasabuvir

A549431-10MG A549431-1MG A549431-250MG
77017-648EA 21.22 USD
77017-648 77017-650 77684-393
N-(6-(3-(tert-Butyl)-5-(2,4-dioxo-3,4-dihydropyrimidin-1(2H)-yl)-2-methoxyphenyl)naphthalen-2-yl)methanesulfonamide 98%, Ambeed.Inc
N-(6-(3-(tert-Butyl)-5-(2,4-dioxo-3,4-dihydropyrimidin-1(2H)-yl)-2-methoxyphenyl)naphthalen-2-yl)methanesulfonamide

Precaución: For research use only.

Formula: C₂₆H₂₇N₃O₅S
MW: 493.57 g/mol
MDL Number: MFCD27923655
CAS Number: 1132935-63-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR