Boc-2,4-dichloro-D-phenylalanine 97%

Supplier: Ambeed
Warning

Synonyms: (R)-2-((tert-Butoxycarbonyl)amino)-3-(2,4-dichlorophenyl)propanoic acid

A548949-1G A548949-250MG A548949-5G A548949-10G
77005-068EA 37.14 USD
77005-068 77005-070 77005-072 77673-462
Boc-2,4-dichloro-D-phenylalanine 97%
Boc-2,4-dichloro-D-phenylalanine

Precaución: for Research Use Only

Formula: C₁₄H₁₇Cl₂NO₄
MW: 334.2 g/mol
MDL Number: MFCD01862941
CAS Number: 114873-12-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR