2-(Bromoethyl)methylsulfone 97%

Supplier: Ambeed
Danger

Synonyms: 1-Bromo-2-(methylsulphonyl)ethane, 2-Bromoethyl Methyl sulphone

A548545-1G A548545-25G A548545-5G
76991-666EA 24.41 USD
76991-666 76991-668 76991-670
2-(Bromoethyl)methylsulfone 97%
2-(Bromoethyl)methylsulfone

Precaución: for Research Use Only

Formula: C₃H₇BrO₂S
MW: 187.055 g/mol
MDL Number: MFCD08274926
CAS Number: 16523-02-7
UN: 1759
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR