{3-[(Tetrahydrofuran-2-ylmethyl)carbamoyl]phenyl}boronic acid pinacol ester 96%

Supplier: Ambeed
Warning

Synonyms: N-((Tetrahydrofuran-2-yl)methyl)-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide

A548317-1G A548317-250MG A548317-5G
77017-642EA 29 USD
77017-642 77017-644 77017-646
{3-[(Tetrahydrofuran-2-ylmethyl)carbamoyl]phenyl}boronic acid pinacol ester 96%
{3-[(Tetrahydrofuran-2-ylmethyl)carbamoyl]phenyl}boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₈H₂₆BNO₄
MDL Number: MFCD09266189
CAS Number: 1073355-06-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR