tert-Butyl-3-(chloromethyl)benzoate 98%

Supplier: Ambeed
Danger

A547883-1G A547883-5G
77034-830EA 23 USD
77034-830 77872-918
tert-Butyl-3-(chloromethyl)benzoate 98%
tert-Butyl-3-(chloromethyl)benzoate

Precaución: for Research Use Only

Formula: C₁₂H₁₅ClO₂
MDL Number: MFCD02180879
CAS Number: 220510-74-7
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR