Ritonavir 98%

Supplier: Ambeed
Warning

Synonyms: A-84538, Thiazol-5-ylmethyl ((2S,3S,5S)-3-hydroxy-5-((S)-2-(3-((2-isopropylthiazol-4-yl)methyl)-3-methylureido)-3-methylbutanamido)-1,6-diphenylhexan-2-yl)carbamate, ABT-538, Abbott 84538

A547820-100G A547820-100MG A547820-10MG A547820-1G A547820-1MG A547820-250MG A547820-50MG A547820-5G A547820-25G
76991-650EA 1822 USD
76991-650 76991-652 76991-654 76991-656 76991-658 76991-660 76991-662 76991-664 77594-674
Ritonavir 98%
Ritonavir

Precaución: for Research Use Only

Formula: C₃₇H₄₈N₆O₅S₂
MDL Number: MFCD00927142
CAS Number: 155213-67-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR