5-Nitro-4-hydroxyquinazoline 96%

Supplier: Ambeed
Warning

A547760-100MG A547760-250MG
77005-064EA 44 USD
77005-064 77005-066
5-Nitro-4-hydroxyquinazoline 96%
5-Nitro-4-hydroxyquinazoline

Precaución: for Research Use Only

Formula: C₈H₅N₃O₃
MDL Number: MFCD12198902
CAS Number: 99768-67-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR