10,15-Dihydro-5H-diindolo[3,2-a:3',2'-c]carbazole 98%

Supplier: Ambeed
Warning

Synonyms: 10,15-Dihydro-5H-5,10,15-triazadiindeno[1,2-a:1',2'-c]fluorene

A547033-1G A547033-250MG A547033-5G
76991-636EA 115.64 USD
76991-636 76991-638 76991-640
10,15-Dihydro-5H-diindolo[3,2-a:3',2'-c]carbazole 98%
10,15-Dihydro-5H-diindolo[3,2-a:3',2'-c]carbazole

Precaución: for Research Use Only

Formula: C₂₄H₁₅N₃
MW: 345.41 g/mol
MDL Number: MFCD21609440
CAS Number: 109005-10-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR