(3-(Dimethylamino)propyl)triphenylphosphonium bromide 95%
Supplier: Ambeed
|
Warning
|
A545311-1G
A545311-250MG
A545311-5G
77034-814EA
38.2
USD
77034-814
77034-816
77034-818
(3-(Dimethylamino)propyl)triphenylphosphonium bromide 95%
(3-(Dimethylamino)propyl)triphenylphosphonium bromide
Precaución: for Research Use Only
Formula:
C₂₃H₂₇BrNP MW: 428.34 g/mol |
MDL Number:
MFCD00012200 CAS Number: 18355-96-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...