2-(4-Methyl-5-thiazolyl)ethyl decanoate 98%

Supplier: Ambeed
Warning

A545296-100G A545296-25G A545296-5G
77034-808EA 111.4 USD
77034-808 77034-810 77034-812
2-(4-Methyl-5-thiazolyl)ethyl decanoate 98%
2-(4-Methyl-5-thiazolyl)ethyl decanoate

Precaución: for Research Use Only

Formula: C₁₆H₂₇NO₂S
MW: 297.46 g/mol
MDL Number: MFCD09032915
CAS Number: 101426-31-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR