(3-(2-Methoxy-2-oxoethoxy)phenyl)boronic acid 95%

Supplier: Ambeed
Warning

A545050-100MG A545050-1G A545050-250MG A545050-5G
77017-618EA 39 USD
77017-618 77017-620 77017-622 77572-732
(3-(2-Methoxy-2-oxoethoxy)phenyl)boronic acid 95%
(3-(2-Methoxy-2-oxoethoxy)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₉H₁₁BO₅
MW: 209.99 g/mol
MDL Number: MFCD11973991
CAS Number: 1256355-55-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR