1-Chloro-3-iodo-2-nitrobenzene 97%

Supplier: Ambeed
Warning

A544912-10G A544912-1G A544912-5G
77005-040EA 190.97 USD
77005-040 77005-042 77005-044
1-Chloro-3-iodo-2-nitrobenzene 97%
1-Chloro-3-iodo-2-nitrobenzene

Precaución: for Research Use Only

Formula: C₆H₃ClINO₂
MW: 283.4510 g/mol
MDL Number: MFCD08443947
CAS Number: 937601-42-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR