(1R,3R)-Methyl 1-(benzo[d][1,3]dioxol-5-yl)-2-(2-chloroacetyl)-2,3,4,9-tetrahydro-1H-pyrido[3,4-b]indole-3-carboxylate 95%

Supplier: Ambeed
Warning

A544621-100G A544621-1G A544621-25G A544621-500G A544621-5G A544621-10G
77005-030EA 252 USD
77005-030 77005-032 77005-034 77005-036 77005-038 77882-320
(1R,3R)-Methyl 1-(benzo[d][1,3]dioxol-5-yl)-2-(2-chloroacetyl)-2,3,4,9-tetrahydro-1H-pyrido[3,4-b]indole-3-carboxylate 95%
(1R,3R)-Methyl 1-(benzo[d][1,3]dioxol-5-yl)-2-(2-chloroacetyl)-2,3,4,9-tetrahydro-1H-pyrido[3,4-b]indole-3-carboxylate

Precaución: for Research Use Only

Formula: C₂₂H₁₉ClN₂O₅
MDL Number: MFCD04973633
CAS Number: 171489-59-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR