4-Chloro-3,5-bis(trifluoromethyl)benzoic acid 95%

Supplier: Ambeed
Warning

A544065-100MG A544065-1G A544065-250MG
76991-608EA 89 USD
76991-608 76991-610 76991-612
4-Chloro-3,5-bis(trifluoromethyl)benzoic acid 95%
4-Chloro-3,5-bis(trifluoromethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₉H₃ClF₆O₂
MDL Number: MFCD06208252
CAS Number: 773108-93-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR