N-Cyclopropyl-2-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)acetamide 96%

Supplier: Ambeed
Warning

A543919-100MG A543919-1G A543919-250MG A543919-5G
77017-610EA 39 USD
77017-610 77017-612 77017-614 77017-616
N-Cyclopropyl-2-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)acetamide 96%
N-Cyclopropyl-2-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)acetamide

Precaución: for Research Use Only

Formula: C₁₇H₂₄BNO₃
MDL Number: MFCD16660305
CAS Number: 1031747-36-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR