Methyl 5-fluorobenzofuran-2-carboxylate 95%

Supplier: Ambeed
Warning

A543743-100MG A543743-1G A543743-250MG A543743-5G
77017-602EA 45.62 USD
77017-602 77017-604 77017-606 77017-608
Methyl 5-fluorobenzofuran-2-carboxylate 95%
Methyl 5-fluorobenzofuran-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₇FO₃
MW: 194.15 g/mol
MDL Number: MFCD08753859
CAS Number: 849236-64-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR