2'-Deoxyadenosine monohydrate 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
(2R,3S,5R)-5-(6-Amino-9H-purin-9-yl)-2-(hydroxymethyl)tetrahydrofuran-3-ol hydrate
A543442-100G
A543442-100MG
A543442-10G
A543442-1G
A543442-25G
A543442-5G
77005-018EA
267.35
USD
77005-018
77005-020
77005-022
77005-024
77005-026
77005-028
2'-Deoxyadenosine monohydrate 95%
2'-Deoxyadenosine monohydrate
Precaución: for Research Use Only
Formula:
C₁₀H₁₃N₅O₃·1H₂O MW: 269.26 g/mol (251.24 anhy) |
MDL Number:
MFCD00149364 CAS Number: 16373-93-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...