4,4'-Diethynylbiphenyl 97%

Supplier: Ambeed
Warning

Synonyms: 4,4'-Diethynyl-1,1'-biphenyl

A543075-100MG A543075-1G A543075-250MG A543075-5G
77017-582EA 13.8 USD
77017-582 77017-584 77017-586 77017-588
4,4'-Diethynylbiphenyl 97%
4,4'-Diethynylbiphenyl

Precaución: for Research Use Only

Formula: C₁₆H₁₀
MW: 202.26 g/mol
MDL Number: MFCD00458964
CAS Number: 38215-38-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR