3-(1H-1,2,4-Triazol-3-yl)pyridine 95%

Supplier: Ambeed
Warning

A540343-100MG A540343-250MG
77017-572EA 42.44 USD
77017-572 77017-574
3-(1H-1,2,4-Triazol-3-yl)pyridine 95%
3-(1H-1,2,4-Triazol-3-yl)pyridine

Precaución: for Research Use Only

Formula: C₇H₆N₄
MDL Number: MFCD18260902
CAS Number: 23195-63-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR