3-Chloro-2,6-difluoropyridine 95%

Supplier: Ambeed
Danger

A540096-1G A540096-250MG A540096-5G A540096-25G
77005-012EA 12 USD
77005-012 77005-014 77005-016 77673-446
3-Chloro-2,6-difluoropyridine 95%
3-Chloro-2,6-difluoropyridine

Precaución: for Research Use Only

Formula: C₅H₂ClF₂N
MDL Number: MFCD11044285
CAS Number: 52208-56-7
UN: 2924
ADR: 3,8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR