(2-(Pyrrolidin-1-yl)pyridin-3-yl)methanamine hydrochloride 95%
Supplier: Ambeed
|
Warning
|
A539074-100MG
A539074-1G
A539074-250MG
77017-558EA
25.47
USD
77017-558
77017-560
77017-562
(2-(Pyrrolidin-1-yl)pyridin-3-yl)methanamine hydrochloride 95%
(2-(Pyrrolidin-1-yl)pyridin-3-yl)methanamine hydrochloride
Precaución: for Research Use Only
Formula:
C₁₀H₁₆ClN₃ MW: 213.71 g/mol |
MDL Number:
MFCD26967957 CAS Number: 1588441-33-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...