(4,5-Difluoro-2-methylphenyl)boronic acid 97%

Supplier: Ambeed
Warning

Synonyms: 4,5-Difluoro-2-methylphenylboronic acid

A538794-1G A538794-250MG A538794-5G
76991-584EA 28 USD
76991-584 77891-089 77891-090
(4,5-Difluoro-2-methylphenyl)boronic acid 97%
(4,5-Difluoro-2-methylphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₇H₇BF₂O₂
MDL Number: MFCD12911676
CAS Number: 1416244-48-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR