2-(2-Ethylhexyl)thiophene 98%
Supplier: Ambeed
|
Warning
|
A538556-1G
A538556-250MG
A538556-5G
A538556-25G
76991-578EA
18
USD
76991-578
76991-580
76991-582
77891-088
2-(2-Ethylhexyl)thiophene 98%
2-(2-Ethylhexyl)thiophene
Precaución: for Research Use Only
Formula:
C₁₂H₂₀S |
MDL Number:
MFCD26743668 CAS Number: 4891-44-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...