2-Bromo-6-fluorobenzamide 98%

Supplier: Ambeed
Warning

A537995-10G A537995-1G
76991-574EA 66.84 USD
76991-574 76991-576
2-Bromo-6-fluorobenzamide 98%
2-Bromo-6-fluorobenzamide

Precaución: for Research Use Only

Formula: C₇H₅BrFNO
MW: 218.0231 g/mol
MDL Number: MFCD07368694
CAS Number: 107485-63-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR