2-(Methylsulfonyl)benzaldehyde 98%

Supplier: Ambeed
Warning

A537767-1G A537767-250MG
77004-998EA 60.48 USD
77004-998 77005-000
2-(Methylsulfonyl)benzaldehyde 98%
2-(Methylsulfonyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₈H₈O₃S
MW: 184.21 g/mol
MDL Number: MFCD11052332
CAS Number: 5395-89-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR