8-Methylquinoline-5-boronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 8-Methyl-5-quinolineboronic Acid

A537553-100MG A537553-1G A537553-250MG A537553-5G
77034-794EA 37 USD
77034-794 77034-796 77034-798 77673-438
8-Methylquinoline-5-boronic acid 98%
8-Methylquinoline-5-boronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₀BNO₂
MW: 187.0000 g/mol
MDL Number: MFCD05256435
CAS Number: 1025010-58-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR