3-Iodo-1H-indazole-6-carboxylic acid 96%

Supplier: Ambeed
Warning

A537442-1G A537442-5G
77017-554EA 14 USD
77017-554 77017-556
3-Iodo-1H-indazole-6-carboxylic acid 96%
3-Iodo-1H-indazole-6-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₅IN₂O₂
MDL Number: MFCD11499016
CAS Number: 1086391-11-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR