2-Methyl-4-nitrobenzene-1-sulfonyl chloride 95%

Supplier: Ambeed
Danger

A537254-1G A537254-5G A537254-250MG
77017-550EA 56 USD
77017-550 77017-552 77613-684
2-Methyl-4-nitrobenzene-1-sulfonyl chloride 95%
2-Methyl-4-nitrobenzene-1-sulfonyl chloride

Precaución: for Research Use Only

Formula: C₇H₆ClNO₄S
MDL Number: MFCD09050619
CAS Number: 21320-90-1
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR