(1-Heptyl)triphenylphosphonium bromide 97%

Supplier: Ambeed
Warning

Synonyms: Heptyltriphenylphosphonium bromide

A537207-25G A537207-500G
77004-986EA 15.92 USD
77004-986 77004-988
(1-Heptyl)triphenylphosphonium bromide 97%
(1-Heptyl)triphenylphosphonium bromide

Precaución: for Research Use Only

Formula: H₃C(CH₂)₆P(C₆H₅)₃Br
MW: 441.39 g/mol
MDL Number: MFCD00050249
CAS Number: 13423-48-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR